Assay Details
| Gene Symbol: RAB21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047559.1 | XP_017903048.1 | 5 - 6 | 701 | 82 | |
| GenBank mRNA | JO439180.2 | - | 5 - 6 | 699 | 82 | |
| JO445863.2 | - | 5 - 6 | 699 | 82 | ||
More Information
Set Membership: |
|

RAB21
Genomic Map