Assay Details
| Gene Symbol: RNF128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013976196.2 | XP_013831650.2 | 6 - 7 | 1410 | 64 | |
| XM_018044229.1 | XP_017899718.1 | 6 - 7 | 1284 | 64 | ||
More Information
Set Membership: |
|

RNF128
Genomic Map