Assay Details
| Gene Symbol: C7H19orf53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051216.1 | XP_017906705.1 | 1 - 2 | 151 | 93 | |
| GenBank mRNA | JO410851.2 | - | 1 - 2 | 161 | 93 | |
| KA350847.1 | - | 1 - 2 | 135 | 93 | ||
More Information
Set Membership: |
|

C7H19orf53
Genomic Map