Assay Details
| Gene Symbol: ETV6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680827.3 | XP_005680884.1 | 7 - 8 | 1524 | 68 | |
| XM_018048473.1 | XP_017903962.1 | 7 - 8 | 1413 | 68 | ||
| GenBank mRNA | JO426832.1 | - | 1550 | 68 | ||
More Information
Set Membership: |
|

ETV6
Genomic Map