Assay Details
| Gene Symbol: DCTN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684082.3 | XP_005684139.1 | 5 - 6 | 475 | 57 | |
| GenBank mRNA | JO576399.1 | - | 5 - 6 | 407 | 57 | |
| JO576401.1 | - | 1 - 2 | 367 | 57 | ||
More Information
Set Membership: |
|

DCTN3
Genomic Map