Assay Details
| Gene Symbol: MOB1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686347.3 | XP_005686404.1 | 5 - 6 | 816 | 72 | |
| GenBank mRNA | JO579496.1 | - | 1 - 2 | 1633 | 72 | |
More Information
Set Membership: |
|

MOB1A
Genomic Map