Assay Details
| Gene Symbol: GSKIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013973150.2 | XP_013828604.1 | 2 - 3 | 404 | 59 | |
| XM_018066472.1 | XP_017921961.1 | 2 - 3 | 329 | 59 | ||
More Information
Set Membership: |
|

GSKIP
Genomic Map