Assay Details
| Gene Symbol: TIMMDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005674956.3 | XP_005675013.2 | 6 - 7 | 946 | 64 | |
| GenBank mRNA | JO336576.2 | - | 490 | 64 | ||
| JO355118.2 | - | 1 - 2 | 268 | 64 | ||
| JO391573.2 | - | 6 - 7 | 563 | 64 | ||
| JO399230.2 | - | 3 - 4 | 218 | 64 | ||
| JO441394.2 | - | 1 - 2 | 387 | 64 | ||
| JO552198.2 | - | 4 - 5 | 302 | 64 | ||
| JO595300.2 | - | 1 - 2 | 385 | 64 | ||
| JO617418.2 | - | 1 - 2 | 375 | 64 | ||
| JT737650.1 | - | 6 - 7 | 653 | 64 | ||
| KA350971.1 | - | 6 - 7 | 961 | 64 | ||
More Information
Set Membership: |
|

TIMMDC1
Genomic Map