Assay Details
| Gene Symbol: LEO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685765.3 | XP_005685822.1 | 8 - 9 | 1557 | 64 | |
| GenBank mRNA | JO429479.2 | - | 4 - 5 | 600 | 64 | |
More Information
Set Membership: |
|

LEO1
Genomic Map