Assay Details
| Gene Symbol: CORO1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061086.1 | XP_017916575.1 | 3 - 4 | 329 | 56 | |
| XM_018061087.1 | XP_017916576.1 | 4 - 5 | 571 | 56 | ||
More Information
Set Membership: |
|

CORO1C
Genomic Map