Assay Details
| Gene Symbol: TMEM38A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013965020.2 | XP_013820474.1 | 3 - 4 | 548 | 75 | |
| GenBank mRNA | JO356679.2 | - | 3 - 4 | 1802 | 75 | |
| JO439328.2 | - | 3 - 4 | 532 | 75 | ||
More Information
Set Membership: |
|

TMEM38A
Genomic Map