Assay Details
| Gene Symbol: ALG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013964097.2 | XP_013819551.1 | 8 - 9 | 1178 | 63 | |
| GenBank mRNA | KA335133.1 | - | 1 - 2 | 320 | 63 | |
More Information
Set Membership: |
|

ALG3
Genomic Map