Assay Details
| Gene Symbol: S100A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677501.2 | XP_005677558.1 | 2 - 3 | 249 | 66 | |
| GenBank mRNA | JT754939.1 | - | 1 - 2 | 96 | 66 | |
| KA347535.1 | - | 1 - 2 | 212 | 66 | ||
More Information
Set Membership: |
|

S100A3
Genomic Map