Assay Details
| Gene Symbol: ARF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679429.3 | XP_005679486.1 | 3 - 4 | 414 | 62 | |
| GenBank mRNA | JO634134.2 | - | 1098 | 62 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: ARF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679429.3 | XP_005679486.1 | 3 - 4 | 414 | 62 | |
| GenBank mRNA | JO634134.2 | - | 1098 | 62 | ||
Set Membership: |
|