Assay Details
| Gene Symbol: SH3RF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683093.3 | XP_005683150.2 | 3 - 4 | 1039 | 75 | |
| XM_013965657.2 | XP_013821111.2 | 2 - 3 | 1262 | 75 | ||
| XM_013965658.2 | XP_013821112.2 | 2 - 3 | 1261 | 75 | ||
| XM_013965659.2 | XP_013821113.2 | 2 - 3 | 1261 | 75 | ||
| XM_013965660.2 | XP_013821114.2 | 2 - 3 | 1262 | 75 | ||
| GenBank mRNA | KA335057.1 | - | 2 - 3 | 776 | 75 | |
More Information
Set Membership: |
|

SH3RF2
Genomic Map