Assay Details
| Gene Symbol: TYRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285727.1 | NP_001272656.1 | 3 - 4 | 821 | 62 | |
| GenBank mRNA | AF136926.2 | - | 3 - 4 | 824 | 62 | |
| HQ391904.1 | - | 2 - 3 | 681 | 62 | ||
| JT760449.1 | - | 4 - 5 | 671 | 62 | ||
More Information
Set Membership: |
|

TYRP1
Genomic Map