Assay Details
| Gene Symbol: CCDC43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693936.3 | XP_005693993.2 | 4 - 5 | 543 | 68 | |
| GenBank mRNA | JO577482.1 | - | 4 - 5 | 539 | 68 | |
More Information
Set Membership: |
|

CCDC43
Genomic Map