Assay Details
| Gene Symbol: FKBP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675910.3 | XP_005675967.1 | 1 - 2 | 371 | 80 | |
| XM_005675911.2 | XP_005675968.1 | 1 - 2 | 288 | 80 | ||
| GenBank mRNA | JO444708.2 | - | 3 - 4 | 560 | 80 | |
More Information
Set Membership: |
|

FKBP7
Genomic Map