Assay Details
| Gene Symbol: ZRANB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698591.3 | XP_005698648.1 | 6 - 7 | 1608 | 72 | |
| XM_018041151.1 | XP_017896640.1 | 6 - 7 | 1567 | 72 | ||
More Information
Set Membership: |
|

ZRANB1
Genomic Map