Assay Details
| Gene Symbol: SNX12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700687.3 | XP_005700744.3 | 1 - 2 | 312 | 73 | |
| GenBank mRNA | JO432071.2 | - | 557 | 73 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: SNX12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700687.3 | XP_005700744.3 | 1 - 2 | 312 | 73 | |
| GenBank mRNA | JO432071.2 | - | 557 | 73 | ||
Set Membership: |
|