Assay Details
Gene Symbol: MCRIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063577.1 | XP_017919066.1 | 4 - 5 | 339 | 71 | |
XM_018063578.1 | XP_017919067.1 | 4 - 5 | 594 | 71 | ||
XM_018063579.1 | XP_017919068.1 | 3 - 4 | 2014 | 71 | ||
GenBank mRNA | JO368961.2 | - | 1 - 2 | 894 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
