Assay Details
| Gene Symbol: WTAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018053429.1 | XP_017908918.1 | 5 - 6 | 471 | 65 | |
| XM_018053430.1 | XP_017908919.1 | 5 - 6 | 1016 | 65 | ||
| XM_018053431.1 | XP_017908920.1 | 5 - 6 | 472 | 65 | ||
| GenBank mRNA | JT685141.1 | - | 6 | 1430 | 65 | |
| KA345319.1 | - | 1023 | 65 | |||
More Information
Set Membership: |
|

WTAP
Genomic Map