Assay Details
| Gene Symbol: TWNK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698333.3 | XP_005698390.1 | 3 - 4 | 2163 | 83 | |
| GenBank mRNA | JO425806.1 | - | 2 - 3 | 1392 | 83 | |
| JO585257.2 | - | 2 - 3 | 1350 | 83 | ||
More Information
Set Membership: |
|

TWNK
Genomic Map