Assay Details
| Gene Symbol: GSK3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062492.1 | XP_017917981.1 | 5 - 6 | 1537 | 53 | |
| GenBank mRNA | JO366011.2 | - | 5 - 6 | 760 | 53 | |
| JO392274.2 | - | 4 - 5 | 490 | 53 | ||
More Information
Set Membership: |
|

GSK3A
Genomic Map