Assay Details
| Gene Symbol: MCUR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039134.1 | XP_017894623.1 | 7 - 8 | 1052 | 69 | |
| GenBank mRNA | JO428573.2 | - | 2 - 3 | 1684 | 69 | |
More Information
Set Membership: |
|

MCUR1
Genomic Map