Assay Details
| Gene Symbol: TCP11L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680573.3 | XP_005680630.1 | 4 - 5 | 604 | 72 | |
| XM_005680574.3 | XP_005680631.1 | 4 - 5 | 517 | 72 | ||
| GenBank mRNA | JO351134.2 | - | 3 - 4 | 529 | 72 | |
| JO416561.2 | - | 3 - 4 | 393 | 72 | ||
More Information
Set Membership: |
|

TCP11L2
Genomic Map