Assay Details
| Gene Symbol: LOC108633295 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044510.1 | XP_017899999.1 | 2611 | 103 | ||
| GenBank mRNA | JO389362.2 | - | 780 | 103 | ||
| JO389368.2 | - | 780 | 103 | |||
More Information
Set Membership: |
|

LOC108633295
Genomic Map