Assay Details
| Gene Symbol: TLCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064316.1 | XP_017919805.1 | 2 - 3 | 1128 | 100 | |
| XM_018064317.1 | XP_017919806.1 | 2 - 3 | 1137 | 100 | ||
| GenBank mRNA | JO440177.2 | - | 2 - 3 | 472 | 100 | |
| KA347549.1 | - | 2 - 3 | 457 | 100 | ||
More Information
Set Membership: |
|

TLCD1
Genomic Map