Assay Details
| Gene Symbol: STYK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680833.3 | XP_005680890.1 | 10 - 11 | 1650 | 60 | |
| XM_005680834.2 | XP_005680891.1 | 7 - 8 | 1074 | 60 | ||
| XM_018048479.1 | XP_017903968.1 | 9 - 10 | 1493 | 60 | ||
| XM_018048480.1 | XP_017903969.1 | 9 - 10 | 1570 | 60 | ||
| XM_018048481.1 | XP_017903970.1 | 9 - 10 | 1372 | 60 | ||
| XM_018048482.1 | XP_017903971.1 | 8 - 9 | 1412 | 60 | ||
More Information
Set Membership: |
|

STYK1
Genomic Map