Assay Details
| Gene Symbol: SHOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048454.1 | XP_017903943.1 | 2 - 3 | 651 | 100 | |
| XM_018048461.1 | XP_017903950.1 | 2 - 3 | 651 | 100 | ||
| XM_018048463.1 | XP_017903952.1 | 3 - 4 | 732 | 100 | ||
| GenBank mRNA | KA341159.1 | - | 3 - 4 | 793 | 100 | |
More Information
Set Membership: |
|

SHOX2
Genomic Map