Assay Details
| Gene Symbol: CDNF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057133.1 | XP_017912622.1 | 3 - 4 | 456 | 76 | |
| GenBank mRNA | JO387324.2 | - | 1 - 2 | 462 | 76 | |
| JT741739.1 | - | 2 - 3 | 526 | 76 | ||
More Information
Set Membership: |
|

CDNF
Genomic Map