Assay Details
| Gene Symbol: TRIM62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058094.1 | XP_017913583.1 | 3 - 4 | 748 | 62 | |
| GenBank mRNA | KA329944.1 | - | 4 - 5 | 1654 | 62 | |
More Information
Set Membership: |
|

TRIM62
Genomic Map