Assay Details
| Gene Symbol: METTL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013974341.2 | XP_013829795.1 | 8 - 9 | 1468 | 76 | |
| XM_013974342.2 | XP_013829796.1 | 8 - 9 | 1643 | 76 | ||
| XM_018039805.1 | XP_017895294.1 | 8 - 9 | 1468 | 76 | ||
| XM_018039806.1 | XP_017895295.1 | 8 - 9 | 1465 | 76 | ||
More Information
Set Membership: |
|

METTL4
Genomic Map