Assay Details
| Gene Symbol: SLC41A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048197.1 | XP_017903686.1 | 8 - 9 | 1665 | 69 | |
| XM_018048198.1 | XP_017903687.1 | 8 - 9 | 1497 | 69 | ||
| XM_018048199.1 | XP_017903688.1 | 8 - 9 | 1528 | 69 | ||
| GenBank mRNA | JO420967.2 | - | 8 - 9 | 1705 | 69 | |
More Information
Set Membership: |
|

SLC41A2
Genomic Map