Assay Details
| Gene Symbol: ZDHHC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043358.1 | XP_017898847.1 | 1 - 2 | 335 | 54 | |
| GenBank mRNA | JO431975.2 | - | 1 - 2 | 323 | 54 | |
| JO608494.2 | - | 1 - 2 | 343 | 54 | ||
More Information
Set Membership: |
|

ZDHHC24
Genomic Map