Assay Details
| Gene Symbol: UBE2R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052369.1 | XP_017907858.1 | 3 - 4 | 892 | 83 | |
| GenBank mRNA | JO425315.1 | - | 3 - 4 | 938 | 83 | |
More Information
Set Membership: |
|

UBE2R2
Genomic Map