Assay Details
| Gene Symbol: HVCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691440.3 | XP_005691497.2 | 4 - 5 | 518 | 64 | |
| XM_018061258.1 | XP_017916747.1 | 4 - 5 | 453 | 64 | ||
| XM_018061260.1 | XP_017916749.1 | 3 - 4 | 676 | 64 | ||
| XM_018061261.1 | XP_017916750.1 | 5 - 6 | 599 | 64 | ||
More Information
Set Membership: |
|

HVCN1
Genomic Map