Assay Details
| Gene Symbol: LOC102191423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013965702.2 | XP_013821156.1 | 10 - 11 | 1194 | 79 | |
| XM_013965703.2 | XP_013821157.1 | 10 - 11 | 1299 | 79 | ||
| XM_013965704.2 | XP_013821158.1 | 12 - 13 | 1496 | 79 | ||
| XM_013965705.2 | XP_013821159.1 | 11 - 12 | 1416 | 79 | ||
| GenBank mRNA | JO378312.2 | - | 2 - 3 | 348 | 79 | |
More Information
Set Membership: |
|

LOC102191423
Genomic Map