Assay Details
| Gene Symbol: TMED4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047499.1 | XP_017902988.1 | 4 - 5 | 557 | 63 | |
| GenBank mRNA | JO433189.2 | - | 3 - 4 | 457 | 63 | |
More Information
Set Membership: |
|

TMED4
Genomic Map