Assay Details
| Gene Symbol: RNF43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064116.1 | XP_017919605.1 | 6 - 7 | 2866 | 75 | |
| XM_018064117.1 | XP_017919606.1 | 7 - 8 | 1813 | 75 | ||
| XM_018064118.1 | XP_017919607.1 | 6 - 7 | 2864 | 75 | ||
| XM_018064119.1 | XP_017919608.1 | 5 - 6 | 3037 | 75 | ||
| GenBank mRNA | KA319211.1 | - | 6 - 7 | 1297 | 75 | |
More Information
Set Membership: |
|

RNF43
Genomic Map