Assay Details
| Gene Symbol: LOC102180474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681471.3 | XP_005681528.2 | 1 - 2 | 178 | 61 | |
| GenBank mRNA | JO405275.2 | - | 4 - 5 | 1592 | 61 | |
| JT622606.1 | - | 1 - 2 | 74 | 61 | ||
More Information
Set Membership: |
|

LOC102180474
Genomic Map