Assay Details
| Gene Symbol: GLYATL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690297.3 | XP_005690354.1 | 2 - 3 | 361 | 81 | |
| XM_013969969.2 | XP_013825423.1 | 2 - 3 | 533 | 81 | ||
| GenBank mRNA | KA344534.1 | - | 4 - 5 | 1160 | 81 | |
More Information
Set Membership: |
|

GLYATL2
Genomic Map