Assay Details
| Gene Symbol: ASB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066429.1 | XP_017921918.1 | 4 - 5 | 941 | 84 | |
| XM_018066430.1 | XP_017921919.1 | 4 - 5 | 949 | 84 | ||
| GenBank mRNA | KA311485.1 | - | 5 - 6 | 588 | 84 | |
More Information
Set Membership: |
|

ASB7
Genomic Map