Assay Details
| Gene Symbol: OPN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042646.1 | XP_017898135.1 | 3 - 4 | 653 | 61 | |
| XM_018042647.1 | XP_017898136.1 | 2 - 3 | 327 | 61 | ||
| XM_018042648.1 | XP_017898137.1 | 2 - 3 | 327 | 61 | ||
More Information
Set Membership: |
|

OPN4
Genomic Map