Assay Details
| Gene Symbol: LYL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051249.1 | XP_017906738.1 | 1 - 2 | 939 | 82 | |
| XM_018051250.1 | XP_017906739.1 | 2 - 3 | 1233 | 82 | ||
More Information
Set Membership: |
|

LYL1
Genomic Map