Assay Details
| Gene Symbol: NNAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057841.1 | XP_017913330.1 | 1 - 2 | 968 | 76 | |
| XM_018057843.1 | XP_017913332.1 | 1 - 2 | 968 | 76 | ||
More Information
Set Membership: |
|

NNAT
Genomic Map