Assay Details
| Gene Symbol: OTC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285729.1 | NP_001272658.1 | 227 | 81 | ||
| GenBank mRNA | GQ148781.1 | - | 227 | 81 | ||
| JO419477.2 | - | 1816 | 81 | |||
| Gene Symbol: OTC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043685.1 | XP_017899174.1 | 2 - 3 | 351 | 81 | |
More Information
Set Membership: |
|

OTC
Genomic Map