Assay Details
| Gene Symbol: KRTAP11-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285767.1 | NP_001272696.1 | 1 | 124 | 90 | |
| GenBank mRNA | AY510113.1 | - | 1 | 124 | 90 | |
| KA317188.1 | - | 1 | 141 | 90 | ||
More Information
Set Membership: |
|

KRTAP11-1
Genomic Map