Assay Details
| Gene Symbol: ACVR2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314169.1 | NP_001301098.1 | 7 - 8 | 959 | 69 | |
| XM_018066622.1 | XP_017922111.1 | 6 - 7 | 1410 | 69 | ||
| XM_018066623.1 | XP_017922112.1 | 6 - 7 | 1239 | 69 | ||
| XM_018066624.1 | XP_017922113.1 | 7 - 8 | 1325 | 69 | ||
| GenBank mRNA | KF723431.1 | - | 7 - 8 | 959 | 69 | |
More Information
Set Membership: |
|

ACVR2B
Genomic Map