Assay Details
| Gene Symbol: IGFBP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314243.1 | NP_001301172.1 | 3 - 4 | 642 | 105 | |
| GenBank mRNA | JO438586.2 | - | 3 - 4 | 850 | 105 | |
| JQ341163.1 | - | 3 - 4 | 642 | 105 | ||
More Information
Set Membership: |
|

IGFBP6
Genomic Map